EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Binding studies of porphyrins to human serum albumin using affinity capillary electrophoresis.
PMID 11504054
Abstract
The present work demonstrates that affinity capillary electrophoresis (ACE) can be employed as a valuable and powerful tool for studying the interactions between porphyrins and proteins in biological and biomedical research, such as the development of porphyrins and related compounds as efficient and selective photosensitizers in the photodynamic therapy of cancers. Binding constants of human serum albumin (HSA) to four biological porphyrins (uroporphyrin I, heptacarboxylporphyrin, coproporphyrin I, protoporphyrin IX), which possess a wide range of hydrophobicity, were estimated by ACE. Based on 1:1 molecular association between these individual porphyrins and HSA, the change of the electrophoretic mobility of HSA as a function of porphyrin concentration in the run buffer was measured and the binding constants were calculated from the slope of the Scatchard plots. The binding constant values were found to be 8.80 +/- 0.51 x 10(4) M(-1), 2.39 +/- 0.16 x 10(5) M(-1), 1.61 +/- 0.11 x 10(6) M(-1), and 9.34 +/- 0.30 x 10(6) M(-1) for uroporphyrin I, heptacarboxylporphyrin, coproporphyrin I, and protoporphyrin IX, respectively, and most of these results are in good agreement with those reported in the literature using conventional methods for binding measurements. Additionally, experimental binding constant data obtained using ACE was found to exhibit very good correlation with theoretical hydrophobicity values calculated using the Rekker's hydrophobic fragmental constant method, thus further supporting the hypothesis that the hydrophobicity of the porphyrin side chains play an important role in governing the hydrophobic interaction of porphyrins with serum proteins such as HSA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U3128
Uroporphyrin I dihydrochloride, ≥95%
C40H38N4O16 · 2HCl