EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Targeting of GLUT6 (formerly GLUT9) and GLUT8 in rat adipose cells.
PMID 11513753
Abstract
The subcellular targeting of the two recently cloned novel mammalian glucose transporters, GLUT6 [previously referred to as GLUT9 [Doege, Bocianski, Joost and Schürmann (2000) Biochem. J. 350, 771-776] and GLUT8, was analysed by expression of haemagglutinin (HA)-epitope-tagged GLUTs in transiently transfected primary rat adipose cells. Similar to HA-GLUT4, both transporters, HA-GLUT6 and HA-GLUT8, were retained in intracellular compartments in non-stimulated cells. In contrast, mutation of the N-terminal dileucine motifs in both constructs led to constitutive expression of the proteins on the plasma membrane. Likewise, when endocytosis was blocked by co-expression of a dominant-negative mutant of the dynamin GTPase, wild-type HA-GLUT6 and HA-GLUT8 accumulated on the cell surface. However, in contrast with HA-GLUT4, no translocation of HA-GLUT6 and HA-GLUT8 to the plasma membrane was observed when the cells were stimulated with insulin, phorbol ester or hyperosmolarity. Thus GLUT6 and GLUT8 appear to recycle in a dynamin-dependent manner between internal membranes and the plasma membrane in rat adipose cells, but are unresponsive to stimuli that induce translocation of GLUT4.