EMAIL THIS PAGE TO A FRIEND
Pest management science
METI-acaricide resistance in Tetranychus urticae does not confer resistance to the naphthoquinones.
PMID 11517730
Abstract
The naphthoquinones and the METI group of compounds act on sites associated with mitochondrial respiration, but METI-resistant strains of two-spotted spider mite from Japan and the UK exhibited no cross-resistance to the naphthoquinones. The potential for developing commercial naphthoquinones therefore remains high.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46438
Tebufenpyrad, PESTANAL®, analytical standard
C18H24ClN3O