EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Isolation and structure of korolkoside, a bis-iridoid glucoside from Lonicera korolkovii.
PMID 11520235
Abstract
A new bis-iridoid glucoside, korolkoside (1), was isolated from Lonicera korolkovii, and its structure and stereochemistry were determined by spectroscopic analysis and chemical derivatization. Korolkoside (1) consists of two secologanin moieties that are connected by an acetal linkage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50741
Secologanin, analytical standard
C17H24O10