EMAIL THIS PAGE TO A FRIEND
Insect biochemistry and molecular biology
8-O-acetylharpagide is not an ecdysteroid agonist.
PMID 11520686
Abstract
We have reinvestigated the activity of 8-O-acetylharpagide, an iridoid glucoside, as an ecdysteroid agonist. Elbrecht et al. (Insect Biochem. Mol. Biol. 26 (1996) 519) isolated a preparation of this compound from Ajuga reptans L. and ascribed ecdysteroid agonist activity on the basis of the induction of an ecdysteroid-like response in Drosophila melanogaster KcO cells, the displacement of [3H]ponasterone A from the Drosophila receptor and the activation of an ecdysteroid-regulated gene in a transactivation assay. We provide evidence that the agonist activity derives from contaminating ecdysteroids; A. reptans is a species rich in ecdysteroids. Purified 8-O-acetylharpagide is not active in the D. melanogaster B(II) cell bioassay, neither as an agonist nor as an antagonist, nor does it displace [3H]ponasterone A from dipteran or lepidopteran ecdysteroid receptor complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00118
8-Acetylharpagide, ≥85% (LC/MS-ELSD)
C17H26O11