EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Molecular cloning and characterization of a novel Jasmonate inducible pathogenesis-related class 10 protein gene, JIOsPR10, from rice (Oryza sativa L.) seedling leaves.
PMID 11527396
Abstract
A novel rice (Oryza sativa L.) gene, homologous to a sorghum pathogenesis-related class 10 protein gene, was cloned from a cDNA library prepared from 2-week-old jasmonic acid-treated rice seedling leaves, and named as JIOsPR10 (jasmonate inducible). JIOsPR10 encoded a 160-amino-acid polypeptide with a predicted molecular mass of 17,173.23 Da and a pI of 5.84. JIOsPR10 was highly similar (77%) to the sorghum PR10 protein, but showed less than 55% similarity with other identified PR10s at the amino acid level. Genomic Southern analyses indicated the presence of related genes in the rice genome. The JIOsPR10 transcript was not detected in the healthy leaves, and was not induced after cut. Further expression analysis revealed that the signaling components of defense/stress pathways, jasmonate, salicylate, and H(2)O(2) significantly up-regulated the JIOsPR10 mRNA over the cut control, whereas two other stress regulators, ethylene and abscisic acid, failed to induce its expression. Interestingly the protein phosphatase (PP) inhibitors, cantharidin, endothall, and okadaic acid, rapidly and potently up-regulated the JIOsPR10 expression, suggesting involvement of the phosphorylation/dephosphorylation events. Additionally, the inducible expression of the JIOsPR10 gene was influenced by light signal(s). Finally, the blast pathogen (Magnaporthe grisea) also specifically elicited the accumulation of JIOsPR10 mRNA in leaves. Induction of the JIOsPR10 gene expression by signaling molecules, PP inhibitors and pathogen attack, strongly indicate a role for this novel gene in rice self-defense/stress response(s).