EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Molecular cloning and functional expression of the guinea pig alpha(1a)-adrenoceptor.
PMID 11527538
Abstract
In the present paper, the cloning and expression of the guinea pig alpha(1A)-adrenoceptor is presented. The nucleotide sequence had an open reading frame of 1401 bp that encoded a 466 amino-acid protein with an estimated molecular mass of approximately 51.5 kDa. When the clone was expressed in Cos-1 cells, specific high-affinity binding of [(3)H]prazosin and [(3)H]tamsulosin was observed. Chloroethylclonidine treatment of membranes slightly decreased the total binding with both radioligands. Binding competition experiments using [(3)H]tamsulosin showed the following potency order: (a) for agonists: oxymetazoline >epinephrine>norepinephrine>methoxamine, and (b) for antagonists: prazosin> or 5-methyl-urapidil=benoxathian>phentolamine>BMY 7378 (8-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4,5]decane-7,9-dione). Photoaffinity labeling using [(125)I-aryl]azido-prazosin revealed a major broad band with a molecular mass between 70 and 80 kDa. The receptor was functional, as evidenced by an epinephrine-increased production of [(3)H]inositol phosphates that was blocked by prazosin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B016
Benoxathian hydrochloride, solid
C19H23NO4S · HCl