EMAIL THIS PAGE TO A FRIEND
Organic letters
Solid-phase synthesis of 1,7-disubstituted-1,3,5-triazepane-2,4-diones.
PMID 11529759
Abstract
[reaction: see text]. The solid-phase synthesis of 1,7-disubstituted-1,3,5-triazepane-2,4-diones from resin-bound amino acids is described. The exhaustive reduction of solid-support bound amides with borane afforded the requisite secondary amines, which following treatment with phenyl isocyanatoformate and cleavage, provided the corresponding triazepane-2,4-diones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
478423
Phenyl isocyanatoformate, 97%
C8H5NO3