EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective aromatic borylation in an inert solvent.
PMID 11529768
Abstract
[reaction: see text]. A protocol for performing Rh catalyzed aromatic borylations in cyclohexane has been devised. Borylation at the 5-position of several 1,3-substituted aromatic species ranging from electron-rich (1,3-(NMe(2))(2)C(6)H(4)) to electron-deficient (1,3-(CF(3))(2)C(6)H(4)) yields the corresponding aryl boronate esters. Veratrole was selectively borylated at the 4-position, thus extending regioselectivity to 1,2-substituted benzenes. Selective borylation at the 3-position of an N-protected pyrrole has also been demonstrated, providing a valuable reagent for cross-coupling reactions in a single step.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140155
1,2-Dimethoxybenzene, ReagentPlus®, 99%
C8H10O2
W379905
1,2-Dimethoxybenzene, ≥99%, FG
C8H10O2