EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Synthesis and characterization of the dimercury(I)-linked compound [PPn]4[(Re7C(CO)21Hg)2]. Oxidative cleavage of the mercury-mercury bond leading to carbidoheptarhenate complexes of mercury(II), including [PPN][Re7C(CO)21Hg(S=C(NME2)2)].
PMID 11531437
Abstract
The reaction of [PPN](3)[Re(7)C(CO)(21)] with Hg(2)(NO(3))(2).2H(2)O in dichloromethane formed the complex [PPN](4)[(Re(7)C(CO)(21)Hg)(2)] ([PPN](4)[1]), isolated in 60% yield. Analogous salts of [1](4-) with [PPh(4)](+) and [NEt(4)](+) were also prepared. The crystal structure of [PPN](4)[1] showed that two carbidoheptarhenate cores are linked by a dimercury(I) unit (d(Hg-Hg) = 2.610(4) A), with each individual mercury atom face-bridging. Oxidative cleavage of the Hg-Hg bond in [1](4-) was effected by 4-bromophenyl disulfide to form [Re(7)C(CO)(21)HgSC(6)H(4)Br](2-) ([4](2-)), by I(2) to form [Re(7)C(CO)(21)HgI](2-) ([5](2-)), and by Br(2) to form [Re(7)C(CO)(21)HgBr](2-) ([6](2-)). Oxidation of [1](4-) by ferrocenium ion (2 equiv) in the presence of tetramethylthiourea resulted in the derivative [Re(7)C(CO)(21)HgSC(NMe(2))(2)](-) ([7](-)). The molecular structure of [PPN][7] was determined by X-ray crystallography. This is the first example of a carbidoheptarhenate-mercury complex with a neutral ligand on mercury, and ligand exchange was demonstrated by displacement with triethylphosphine. Complex [7](-) can also be prepared by protonating [Re(7)C(CO)(21)HgO(2)CCH(3)](2-) in the presence of tetramethylthiourea. Cyclic voltammetry data to calibrate and compare the redox properties of compounds [1](4-) and [7](-) have been measured.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
230405
Mercury(I) chloride, ACS reagent, ≥99.5%
Cl2Hg2