EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Resonance Rayleigh scattering study of the interaction of heparin with some basic diphenyl naphthylmethane dyes.
PMID 11534715
Abstract
In a near-neutral medium, a combination of heparin with some basic diphenyl naphthylmethane dyes such as victoria blue 4R (VB4R), victoria blue B (VBB), or night blue (NB) can result in a significant enhancement of resonance. Rayleigh scattering (RRS) and their maximum scattering wavelengths lambdamax appear at 523, 534, and 540 nm for VB4R, VBB, and NB, respectively. The characteristics of RRS spectra of the heparin-dye complexes, the influencing factors, and the optimum conditions of these reactions have been investigated. The RRS intensity is directly proportional to the concentration of heparin in the range of approximately 0 to 0.4 microg/mL for all systems. The methods exhibit high sensitivities, and the detection limits for heparin are 3.35 ng/mL for the VB4R system, 6.62 ng/ mL for the VBB system, and 6.29 ng/mL for the NB system. Because the VB4R system is the most sensitive, it was taken as an example to study the selectivity of the method. A new method for the determination of trace amounts of heparin based on RRS technique has been developed. Moreover, the enhancement reasons of RRS and the relationship between RRS spectral characteristics of the heparin-dye complex and its absorption spectra have been primarily discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94890
Victoria Blue B, for microscopy (Bact., Hist.)
C33H32ClN3