EMAIL THIS PAGE TO A FRIEND
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life
Phosphorylation of glyceric acid in aqueous solution using trimetaphosphate.
PMID 11536746
Abstract
The phosphorylation of glyceric acid is an interesting prebiotic reaction because it converts a simple, potentially prebiotic organic molecule into phosphate derivatives that are central to carbohydrate metabolism. We find that 0.05 M glyceric acid in the presence of 0.5 M trimetaphosphate in alkaline solution gives a mixture of 2- and 3-phosphoglyceric acids in combined yields of up to 40%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8877
D-(−)-3-Phosphoglyceric acid disodium salt, ≥93%, powder
C3H5Na2O7P