EMAIL THIS PAGE TO A FRIEND
The Biological bulletin
Gravitaxis in flagellates.
PMID 11536789
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130079
Methyltriphenylphosphonium bromide, 98%
C19H18BrP