EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Glycolonitrile oligomerization: structure of isolated oxazolines, potential heterocycles on the early earth.
PMID 11543606
Abstract
A study of glycolonitrile polymerization has led to the isolation and characterization of two 2,5-dihydro-4-aminooxazoles, 4 and 5. Previous reports have misassigned these structures as s-triazines or pyrimidines. X-ray diffraction analysis of crystals of 4 and an acetylated oxazole derivative of 5 (6) confirm the proposed structures. Ab initio computations are used to assess the relative thermodynamic stability of three trimer isomers (an s-triazine, an aminohydroxypyrimidine, and an aminooxazoline), and the results indicate that 4 is a novel kinetic product. Mechanistic considerations rationalize kinetic oxazole formation over the more customary triazine or pyrimidine trimers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50640
Glycolic acid nitrile solution, ~70% in H2O
C2H3NO