EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Cardiomyocyte-transistor-hybrids for sensor application.
PMID 11544050
Abstract
An extracellular recording system has been designed for the detection of electrical cell signals using p-channel or n-channel field-effect transistor (FET) arrays with non-metallized gates. Signals from rat heart muscle cell were recorded by these devices and the results described on the basis of an equivalent circuit. This technique is sensitive enough to detect minute changes of the extracellular membrane voltage and has potential applications in drug screening. We show that known cardiac stimulants (isoproterenol, norepinephrine) and relaxants (verapamil, carbamylcholine) have characteristic effects on the heart cells in terms of the changes of beat frequencies in the absence or presence of corresponding agents.