EMAIL THIS PAGE TO A FRIEND
The Journal of infection
Disorders of lipid metabolism in patients with HIV disease treated with antiretroviral agents: frequency, relationship with administered drugs, and role of hypolipidaemic therapy with bezafibrate.
PMID 11545549
Abstract
To assess the correlation between antiretroviral treatment and dyslipidaemia in HIV-infected patients, and the role of bezafibrate as a lipid-lowering agent. We retrospectively compared serum lipid levels of five groups of 40 patients, each of them treated with either saquinavir hard gel, indinavir, or ritonavir (associated with two nucleoside analogues), or dual nucleoside reverse transcriptase inhibitors (NRTI) with or without a non-nucleoside reverse transcriptase inhibitor (NNRTI), or not treated with antiretrovirals, randomly selected from nearly 1000 HIV-infected patients followed-up for >or= 12 months, while on the relevant therapy. Hypertriglyceridaemia was defined by triglyceride levels >or= 172 mg/dl, and hypercholesterolaemia by cholesterol levels >or= 200 mg/dl. All patients with triglyceridaemia > 300 mg/dl and cholesterolaemia > 220 mg/dl for at least 6 months, and unresponsive to a >or= 3-month diet, started bezafibrate (400 mg/day), and were prospectively followed-up at a