EMAIL THIS PAGE TO A FRIEND
Der Anaesthesist
[Acute normovolemic hemodilution (ANH). Der Anaesthesist (2000) 49: 939-948].
PMID 11556172
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7780
Propionylpromazine hydrochloride
C20H24N2OS · HCl
46689
Propionylpromazine hydrochloride, VETRANAL, analytical standard
C20H24N2OS · HCl