EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Cloning of a new aquaporin (AQP10) abundantly expressed in duodenum and jejunum.
PMID 11573934
Abstract
A new aquaporin (AQP10) was identified in human small intestine. This gene encoded a 264-amino-acid protein with high sequence identity with AQP3 (53%), 9 (52%), and 7 (43%). These AQPs constitute one subfamily of AQP family that is differentiated from the other subfamily of AQP (AQP0, 1, 2, 4, 5, 6, and 8) by sequence homology. Ribonuclease protection assay and Northern blotting demonstrated almost exclusive expression of AQP10 mRNA in the duodenum and jejunum. In situ hybridization localized it in absorptive jejunal epithelial cells. Xenopus oocytes expressing AQP10 exhibited an increased osmotic water permeability in a mercury-sensitive manner. Although AQP10 belongs to the AQP subfamily, which has been characterized by permeability to water and neutral solutes such as urea and glycerol, it was not permeable to urea nor glycerol. The specific expression of AQP10 suggests its contribution to the water transport in the upper portion of small intestine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2103514
Anti-AQP10 antibody produced in rabbit, affinity isolated antibody