EMAIL THIS PAGE TO A FRIEND
Organic letters
Improved synthesis of C-terminal peptide thioesters on "safety-catch" resins using LiBr/THF.
PMID 11574025
Abstract
[reaction: see text] The alkanesulfonamide "safety-catch" resin has proven useful for Fmoc-based synthesis of C-terminal peptide thioesters. We now report that the yield of isolated thioester can increase significantly when the cleavage reaction is carried out in 2 M LiBr/THF rather than DMF or THF. The largest effects are seen with problematic peptides that aggregate or form secondary structures on the resin.