EMAIL THIS PAGE TO A FRIEND
Current opinion in investigational drugs (London, England : 2000)
Donitriptan (Pierre Fabre).
PMID 11575714
Abstract
Pierre Fabre is developing donitriptan, a piperazide 5-HT1D agonist, as a potential treatment for migraine [175854]. In January 2001, donitriptan had completed phase I trials for migraine and was scheduled to enter phase II development [396027]. This compound has an increased potency and, importantly, markedly higher intrinsic activity in comparison to the well described tryptamine derivatives, naratriptan, zolmitriptan and sumatriptan [295769].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9071
Donitriptan monohydrochloride, ≥98% (HPLC)
C23H25N5O2 · HCl