EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Plasticisation of amylodextrin by moisture. Consequences for compaction behaviour and tablet properties.
PMID 11576830
Abstract
Amylodextrin, a starch-based controlled release excipient, spontaneously absorbs moisture during storage. The aim of this study was to investigate plasticisation of amylodextrin by moisture and its effect on compaction and tablet characteristics. The glass transition temperature (T(g)) of amylodextrin powders with moisture fractions (x(w)) 0.070
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9765
Starch, soluble, ACS reagent