EMAIL THIS PAGE TO A FRIEND
Chemosphere
Seasonal patterns of non-terpenoid C6-C10 VOC emission from seven Mediterranean woody species.
PMID 11592412
Abstract
The seasonal pattern of non-terpenoid C6-C10 VOC emission by seven Mediterranean woody species (Bupleurum fruticosum, Cistus albidus, Pinus halepensis, Arbutus unedo, Erica arborea, Quercus coccifera, and Q. ilex) was studied under field conditions. Branch chamber samples were sorbed on carbotrap and analyzed by thermal desorption in combination with GC-MS. These non-terpenoid C6-C10 VOC emissions were large, almost of similar magnitude to those of terpenes. Overall, maximum values were recorded in spring and summer (up to 12 microg g(-1) DM h(-1) in Q. ilex) and minimum values in autumn and winter (up to 5 microg g(-1) DM h(-1) in Q. ilex). These C6-C10 VOC emissions represented 2.82% of the photosynthetic C fixation in summer and 0.22% in winter. Some compounds such as 2-ethoxyethyl acetate were emitted by most species, others such as 3-hexen-1-ol, phenol or decanal were significantly emitted only by few species. The greatest diversity of emitted non-terpenoid C6-C10 VOCs was observed in spring and in Q. ilex. Temperature seemed a strong driver of these seasonal changes but other species-specific and seasonal factors seem involved. These results indicate that C6-C10 non-terpenoid VOCs contribute a rather significant fraction of the total biogenic VOC flux from these Mediterranean species, especially in spring and summer, and therefore should be considered in VOC emission inventories and in model predictions of tropospheric chemistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25087
Carbotrap® 100, glass TD tube, preconditioned
20242 Carbotrap® 200 Dynatherm 890, 850, for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16
20865 Carbotrap® 201, for use with Dynatherm ACEM 900/901-FF, I.D. × O.D. × L 1 mm × 6 mm × 7.25 in.
20083-U
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29745-U
Carbotrap® 202, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20013-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29531-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30138-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2 , unconditioned, pkg of 10 ea
30137-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2 , unconditioned, pkg of 10 ea
29744-U
Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29750-U Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20875
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, pkg of 1 ea
28283-U
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
28273-U
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
20370-U Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4  in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
21705-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, preconditioned, pkg of 10 ea
29532-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30141-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., unconditioned, pkg of 10 ea
30139-U
Carbotrap® 300, stainless steel TD tube, unconditioned
29746-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29751-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20881 Carbotrap® 317, glass TD tube (fast-flow), preconditioned
28311-U Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
29752-U Carbotrap® 349, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20287 Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
11325-U Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, pack of 144 × 190 mg
20309 Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
11047-U Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 500 g
10435-U Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
10460-U Carbotrap® Adsorbent, matrix Carbotrap® Y, 20-40 mesh, bottle of 10 g