EMAIL THIS PAGE TO A FRIEND
Biotechnology and applied biochemistry
Production of dextran from sucrose by a newly isolated strain of Leuconostoc mesenteroides (PCSIR-3) with reference to L. mesenteroides NRRL B-512F.
PMID 11592914
Abstract
A newly isolated strain of Leuconostoc mesenteroides (PCSIR-3) produced a different dextran compared with that of L. mesenteroides NRRL B-512F. Different media compositions used for dextran production showed that media containing CaCl(2) produced dextran in higher quantities compared with other media. The viscosity of the dextran produced in different media varied in nature. Dextran from media 1 and 2 was of higher molecular mass compared with that from media 3, 4 and 5. Dextran production is also effected by the sucrose concentration in the media. The higher the initial concentration of sucrose, the higher is the yield of dextran produced per unit volume; however, the percentage conversion of sucrose into dextran decreases. A continuous drop in pH was associated with growth and dextran production. The yield of dextran increases during the growth phase and maximum yield was obtained at the end of the exponential phase. Dextran produced by L. mesenteroides PCSIR-3 is quite different from the dextran produced by NRRL B-512F. Maximum dextran production from L. mesenteroides PCSIR-3 occurs in 18 h compared with 12 h for NRRL B-512F.
Related Materials