EMAIL THIS PAGE TO A FRIEND
Organic letters
A new fluorescent chemosensor for copper ions based on tripeptide glycyl-histidyl-lysine (GHK).
PMID 11594813
Abstract
[structure: see text]. A new fluorescent chemosensor for Cu2+ ions was synthesized by modifying the tripeptide glycyl-histidyl-lysine (GHK) with 9-carbonylanthracene via the standard Fmoc solid-phase peptide synthesis method. While significant fluorescence quenching was observed from the molecule upon binding with Cu2+, addition of Fe2+, Co2+, Ni2+, and Zn2+ to the peptide solution caused a minimum fluorescence emission spectral change, indicating a high specificity of this chemosensor for Cu2+ ions. Effects of pH were also investigated.
Related Materials