EMAIL THIS PAGE TO A FRIEND
Organic letters
A convenient method for 3-pyrroline synthesis.
PMID 11594838
Abstract
[reaction: see text]. The synthesis of a range of 3-pyrrolines has been achieved from primary amine starting materials using a two-step alkylation/alkylidene carbene CH-insertion reaction sequence. We have shown that insertion into a range of CH-bond types is possible, and the formation of nitrogen-bearing quaternary stereocenters is a relatively facile process. The insertion reaction occurs with >95% retention of stereochemistry, but the presence of protecting groups on nitrogen is generally deleterious to the cyclization process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377112
3-Pyrroline, 95%
C4H7N