EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
Cantharidin revisited: a blistering defense of an ancient medicine.
PMID 11594862
Abstract
Cantharidin, a vesicant produced by beetles in the order Coleoptera, has a long history in both folk and traditional medicine. In dermatology, topical cantharidin has long been used to treat warts and molluscum. In 1962, cantharidin lost Food and Drug Administration (FDA) approval owing to the failure of its manufacturers to submit data attesting to cantharidin's efficacy. However, it is expected that the FDA will soon include cantharidin on its "Bulk Substances List," which would permit physicians or pharmacists to compound cantharidin to be used in the office for individual patients. A comprehensive discussion of the origins, folk uses, current FDA status, current dermatologic uses, and effects of cantharidin poisoning has been compiled herein. No cases of systemic intoxication or scarring have been reported with the proper use of cantharidin by a physician. Cantharidin is a safe and valuable medication and should be readded to the dermatologic therapeutic armamentarium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7632
Cantharidin
C10H12O4
SML1537 Cantharidin Ready Made Solution, 0.1 mg/mL in DMSO
C10H12O4