EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
A Korean girl with alpha-aminoadipic and alpha-ketoadipic aciduria accompanied with elevation of 2-hydroxyglutarate and glutarate.
PMID 11596655
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75447
2-Oxoadipic acid, ≥95.0% (HPLC)
C6H8O5