EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
A platinum(IV) complex oxidizes guanine to 8-oxo-guanine in DNA and RNA.
PMID 11599941
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
404322
Dichloro(1,2-diaminocyclohexane)platinum(II)
C6H14Cl2N2Pt