EMAIL THIS PAGE TO A FRIEND
Clinical obstetrics and gynecology
Unconventional estrogens: estriol, biest, and triest.
PMID 11600867
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1680000
Estriol, European Pharmacopoeia (EP) Reference Standard
C18H24O3