EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Nutrition
Chocolate: food as medicine/medicine as food.
PMID 11603654
Abstract
Cocoa and chocolate products have been delicacies for hundreds of years. Only recently have they been recognized as significant sources of phytochemicals with healthful effects. These foods are among the most concentrated sources of the procyanidin flavonoids, catechin and epicatechin. Recent studies have shown that these polyphenols are absorbed from the intestine of animals and humans with epicatechin absorbed much more than catechin. These flavonoids have potent antioxidant and antiplatelet activities following consumption of cocoa or chocolate.