EMAIL THIS PAGE TO A FRIEND
Molecular and biochemical parasitology
Puromycin-N-acetyltransferase as a selectable marker for use in Plasmodium falciparum.
PMID 11606225
Abstract
The limited number of selectable markers available for malaria transfection has hindered extensive manipulation of the Plasmodium falciparum genome and subsequently thorough genetic analysis of this organism. In this paper, we demonstrate that P. falciparum is highly sensitive to the drug puromycin, but that transgenic expression of the puromycin-N-acetyltransferase (PAC) gene from Streptomyces alboninger confers resistance to this drug with the IC(50) and IC(90) values increasing approximately 3- and 7-fold, respectively in PAC-expressing parasites. Despite this relatively low level of resistance, parasite populations transfected with the PAC selectable marker and selected directly on puromycin emerged at the same rate post-transfection as human dihydrofolate reductase (hDHFR)-expressing parasites, selected independently with the anti-folate drug WR99210. Transfected parasites generally maintained the PAC expression plasmid episomally at between two and six copies per parasite. We also demonstrate by cycling transfected parasites in the presence and absence of puromycin for several weeks, that the PAC selectable marker can be used for gene-targeting. Since the mode of action of puromycin is distinct from other drugs currently used for the stable transfection of P. falciparum, the PAC selectable marker should also have applicability for use in conjunction with other positive selectable markers, thereby increasing the possibilities for more complex functional studies of this organism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W1770
WR99210
C14H18Cl3N5O2