EMAIL THIS PAGE TO A FRIEND
Biophysical journal
Cholesterol monohydrate nucleation in ultrathin films on water.
PMID 11606285
Abstract
The growth of a cholesterol crystalline phase, three molecular layers thick at the air-water interface, was monitored by grazing incidence x-ray diffraction and x-ray reflectivity. Upon compression, a cholesterol film transforms from a monolayer of trigonal symmetry and low crystallinity to a trilayer, composed of a highly crystalline bilayer in a rectangular lattice and a disordered top cholesterol layer. This system undergoes a phase transition into a crystalline trilayer incorporating ordered water between the hydroxyl groups of the top and middle sterol layers in an arrangement akin to the triclinic 3-D crystal structure of cholesterol x H(2)O. By comparison, the cholesterol derivative stigmasterol transforms, upon compression, directly into a crystalline trilayer in the rectangular lattice. These results may contribute to an understanding of the onset of cholesterol crystallization in pathological lipid deposits.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151114
Cholesteryl acetate, 97%
C29H48O2