EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Reproducible measurement of single-molecule conductivity.
PMID 11641492
Abstract
A reliable method has been developed for making through-bond electrical contacts to molecules. Current-voltage curves are quantized as integer multiples of one fundamental curve, an observation used to identify single-molecule contacts. The resistance of a single octanedithiol molecule was 900 +/- 50 megohms, based on measurements on more than 1000 single molecules. In contrast, nonbonded contacts to octanethiol monolayers were at least four orders of magnitude more resistive, less reproducible, and had a different voltage dependence, demonstrating that the measurement of intrinsic molecular properties requires chemically bonded contacts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471836
1-Octanethiol, ≥98.5%
C8H18S