EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A Convenient Method for the Preparation of (Z)-alpha,beta-Unsaturated Carbonyl Compounds.
PMID 11667136
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399191
Rhodium(II) trifluoroacetate dimer
C8F12O8Rh2