EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A new approach for dynamics of enzyme-catalyzed glutathione conjugation by electrospray quadrupole/time-of-flight mass spectrometry.
PMID 11673899
Abstract
The dynamics of enzyme-catalyzed glutathione conjugation was studied by electrospray quadrupole/time-of-flight (Q-TOF) mass spectrometry with a nanospray interface. After incubation of human glutathione S-transferase A1-1 (GT) with glutathione (GSH) and an electrophilic substrate, electrospray indicated the presence of enzyme/product adducts such as [2GT + product], [2GT + GSH' + product], and [2GT + 2 products] as well as [2GT] and [2GT + GSH']. The relative abundance of GT/product adduct ions increased with incubation time. The wide m/z range of detection (m/z 300-5000) allowed the observation of product, suggested to be released from enzyme/product adducts, in the same mass spectrum. The noncovalent complexes of GT/product were completely replaced by GT/inhibitor complexes following the addition of GT inhibitor to the incubation mixture. Furthermore, a collision-activated decomposition analysis of these ion species provided us with useful information to interpret or identify ion species. The results suggest that electrospray Q-TOF mass spectrometry is a powerful approach for studying the dynamics of the enzyme reaction as well as the structure of enzyme complexes at high sensitivity.
Related Materials