EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Enzyme-assisted preparation of D-tert.-leucine.
PMID 11676008
Abstract
Optically pure D-tert.-leucine was obtained by the enzymatic hydrolysis of (+/-)-N-acetyl-tert. leucine chloroethyl ester after about 50% conversion, this being catalyzed by a protease from Bacillus licheniformis (Alcalase), and subsequent acidic saponification of the recovered ester. Among the methyl, ethyl, octyl, chloroethyl and trichloroethyl esters, the chloroethyl ester exhibited the highest rate of hydrolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
332178
DL-tert-Leucine, 98%
C6H13NO2
269107
L-tert-Leucine, 99%
C6H13NO2