EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Reproducible fabrication of miniaturized glucose sensors: preparation of sensing membranes for continuous monitoring.
PMID 11679292
Abstract
The immobilization process of glucose oxidase(GOx) in the poly(1,3-diaminobenzene) (poly(1,3-DAB)) network was closely investigated in situ using an electrochemical quartz crystal microbalance(EQCM). GOx captured in approximately 50 nm thick poly-1,3-DAB layer causes a 514 Hz frequency increase, corresponding to 541 ng, and distributes mostly in the outer part of the polymer film. The presence of poly-L-lysine and glutaraldehyde during electropolymerization of poly(1,3-DAB) improves sensitivity by raising the amount of GOx immobilized. Adding a protective membrane on to the enzyme layer from poly(tetrafluoroethylene) (PTFE) dispersed in aqueous media lets the entire fabrication procedure finish perfectly without nonaqueous solvent. The finalized needle-type glucose sensors show competent functions in sensitivity, stability, biocompatibility, lifetime, interference and reproducibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P23954
m-Phenylenediamine, flakes, 99%
C6H8N2