EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Synthesis and bioactivity of natural and C-3 fluorinated biosynthetic precursors of 28-homobrassinolide.
PMID 11684198
Abstract
In this paper we describe the synthesis of two new fluorinated brassinosteroids: (22R,23R)-22,23-dihydroxy-3alpha-fluorostigmastan-6-one and (22R,23R)-22,23-dihydroxy-3beta-fluorostigmastan-6-one. Their bioactivities were evaluated in the rice lamina inclination test and compared with that of 28-homocastasterone, 28-homotyphasterol and 28-homoteasterone, possible biosynthetic precursors of 28-homobrassinolide. Results confirmed expected similarities between the biosynthesis of 24-ethylbrassinosteroids (named as the 28-homo series) and that described for 24-methylbrassinosteroids, and also indicated that these precursors might exhibit per se activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1267
22(S),23(S)-Homobrassinolide
C29H50O6