EMAIL THIS PAGE TO A FRIEND
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993)
[Anti-corrosive effect of pesticides in soil corrosion conditions].
PMID 11692682
Abstract
Effect of some sub-standard pesticides (Ramrod, Linuron, Simazin) with respect to corrosion-active groups of microorganisms: sulphate-reducing bacteria (SRB), denitrifying bacteria (DNB), saprophytic bacteria (SB) and their inhibiting properties under the conditions of active corrosion have been studied to estimate a possibility to use them as biocide additions when producing protective materials. It has been shown that the sub-standard pesticides Ramrod and Simazin are promising for to be used as the biocides additions under the protection of bioresistant materials. It is supposed that inhibitors-biocides may be found in a series of compounds obtained under chemical modification of substandard pesticides Ramrod and Simazin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45637
Propachlor, PESTANAL®, analytical standard
C11H14ClNO