EMAIL THIS PAGE TO A FRIEND
Toxicology and industrial health
Influence of 2-bromopropane on reproductive system--short-term administration of 2-bromopropane inhibits ovulation in F344 rats.
PMID 11693945
Abstract
The present study was performed to investigate the toxic effects of 2-bromopropane (2BP) on the female reproductive system. Female F344 rats were administered 2BP (500 or 1000 mg/kg, i.p.) at intervals of 2 or 3 days for 15-17 days. The body weights were measured and estrous stages were observed throughout the experimental period. Ovulation, organ weights, ovarian histology, and blood biochemistry were investigated on the terminal day of the experiment. Uterine weights in rats treated with 2BP were significantly lower than those in control animals. Body, liver, kidney, and adrenal weights in 2BP-treated rats showed no significant differences from control values. 2BP treatment prolonged estrous cycles and decreased the number of ovulated ova in spontaneous ovulation. In addition, histological examinations showed that the preovulatory follicles in the ovary were altered markedly in 2BP groups. These results show that even in short-term treatment, 2BP injured the ovary, particularly the preovulatory follicles. It appears that these damages of the preovulatory follicles induced by 2BP reduced the numbers of spontaneously ovulated ova in female F344 rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B78114
2-Bromopropane, 99%
C3H7Br
59370
2-Bromopropane, purum, ≥99.0% (GC)
C3H7Br