EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Semiconductor-mediated photocatalysed degradation of two selected pesticide derivatives, terbacil and 2,4,5-tribromoimidazole, in aqueous suspension.
PMID 11695479
Abstract
The photocatalysed degradation of two selected pesticide derivatives, namely 3-tert-butyl-5-chloro-6-methyluracil (terbacil) and 2,4,5-tribromoimidazole (TBI) has been investigated in aqueous suspensions of titanium dioxide (TiO2) under a variety of conditions employing a pH-stat technique. The degradation was studied by monitoring the change in substrate concentration of the model compound employing HPLC analysis, and the decrease in total organic carbon (TOC) content, respectively, as a function of irradiation time. The degradation kinetics were studied under different conditions such as reaction pH, substrate and photocatalyst concentration, type of TiO2 photocatalyst and the presence of alternative additives such as H2O2, KBrO3 and (NH4)2S2O8 besides molecular oxygen. The degradation rates and the photonic efficiencies were found to be strongly influenced by the above parameters. Toxicity tests for the irradiated samples of the pesticide derivatives measuring the luminescence of bacteria Vibrio fischeri after 30 minutes of incubation were also performed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
141704
2,4,5-Tribromoimidazole, 97%
C3HBr3N2
45675
Terbacil, PESTANAL®, analytical standard
C9H13ClN2O2