EMAIL THIS PAGE TO A FRIEND
Phytochemical analysis : PCA
Phenylpropanoid interconversion in Anigozanthos preissii observed by high-performance liquid chromatography-nuclear magnetic resonance spectroscopy.
PMID 11704960
Abstract
HPLC-NMR spectroscopy was employed to observe isotopically labelled intermediates of the phenylpropanoid metabolism in Anigozanthos preissii. Hydroxylation of dihydrocinnamic acid to dihydro-p-coumaric acid and the reversible interconversion between phenylpropanoids and phenylpropenoids were detected, suggesting the operation of a metabolic network in early phenylpropanoid biosynthesis in this plant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H52406
3-(4-Hydroxyphenyl)propionic acid, 98%
C9H10O3