EMAIL THIS PAGE TO A FRIEND
Phytochemical analysis : PCA
NOESY on neurotoxins: NMR and conformational assignments of picrotoxins.
PMID 11704965
Abstract
Nuclear Overhauser effect spectroscopy (NOESY) gave full assignments of the 1H-NMR spectra of the picrotoxane neurotoxins tutin, hyenanchin, picrotoxinin and picrotin, as well as the solution conformations of these compounds, consistent with molecular modelling. Fully assigned 13C-NMR data are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8390
Picrotoxinin
C15H16O6