EMAIL THIS PAGE TO A FRIEND
Phytochemical analysis : PCA
Quantitative determination of liriodenine and moupinamide in five species of Mollinedia by high performance liquid chromatography.
PMID 11708299
Abstract
A simple method for the resolution and subsequent quantitative determination of liriodenine and moupinamide in Mollinedia species was developed using reversed-phase HPLC based on an octadecyl silane-packed column eluted with gradients of methanol and water.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00196
Moupinamide, ≥95% (LC/MS-ELSD)
C18H19NO4