EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Structural and rheological properties of methacrylamide modified gelatin hydrogels.
PMID 11709840
Abstract
Dynamic shear oscillation measurements at small strain were used to characterize the viscoelastic properties and related differences in the molecular structure of hydrogels based on gelatin methacrylamide. Gelatin was derivatized with methacrylamide side groups and was subsequently cross-linked by radical polymerization via photoinitiation. The light treatment of methacrylamide gelatin solutions resulted in the production of hydrogel films with high storage modulus (G'). Mechanical spectra and thermal scanning rheology of the obtained hydrogels are described. The temperature scan of the network below and above melting point of gelatin allowed us to identify the respective contributions of chemical and physical cross-linkage to the hydrogel elastic modulus. The results indicate that the rheological properties of the gelatin-based hydrogels can be controlled by the degree of substitution, polymer concentration, initiator concentration, and UV irradiation conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276685
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, 94%
C8H10O3