EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Molecular recognition in a propazine-imprinted polymer and its application to the determination of triazines in environmental samples.
PMID 11721910
Abstract
An analytical methodology for the determination of triazines in environmental samples incorporating a molecularly imprinted solid-phase extraction (MISPE) process using a propazine-imprinted polymer was developed. Two different polymers were prepared using acetonitrile or toluene as porogen, and their optimum loading, washing, and elution conditions were established. Although both polymers were able to recognize several chlorotriazines (propazine, atrazine, simazine, desethylatrazine, and desisopropylatrazine), the polymer prepared in toluene showed the best performance and was also capable of recognizing a methylthiotriazine (prometryn). A binding study carried out in this polymer demonstrated that it possesses heterogeneous binding sites with different binding abilities. From this study, it was also concluded that desethylatrazine and desisopropylatrazine displace the other triazines at high concentrations, including the template molecule. The accuracy and selectivity of the MISPE process developed was verified using a certified reference material for drinking water containing atrazine and simazine among other commonly used pesticides. Finally, the MISPE procedure was successfully applied to the cleanup of drinking and groundwater, soil, and corn sample extracts, and the triazines were determined by micellar electrokinetic chromatography.