EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Silica-supported TEMPO catalysts: synthesis and application in the Anelli oxidation of alcohols.
PMID 11722219
Abstract
The application of silica-supported TEMPO as a recyclable catalyst in the Anelli oxidation of alcohols is reported. The catalyst is easily obtained in a one-step reductive amination procedure starting from a commercially available aminopropyl-functionalized silica. Details of the synthesis of the supported catalyst and its analysis by MAS NMR are presented. Various alcohol oxidations according to the Anelli protocol have been carried out and the stability of the applied silica-supported TEMPO has been studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
709859
TEMPO on silica gel