EMAIL THIS PAGE TO A FRIEND
Doklady. Biochemistry and biophysics
The antimetastatic and immunomodulating activities of ginseng minor glycosides.
PMID 11727551
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
65839
Ginsenoside Rf, analytical standard
C42H72O14
01580590
Ginsenoside Rf, primary reference standard
C42H72O14