EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Densin-180 interacts with delta-catenin/neural plakophilin-related armadillo repeat protein at synapses.
PMID 11729199
Abstract
Densin-180, a protein purified from the postsynaptic density fraction of the rat forebrain, is the founding member of a newly described family of proteins termed the LAP (leucine-rich repeats and PSD-95/Dlg-A/ZO-1 (PDZ) domains) family that plays essential roles in establishment of cell polarity. To identify Densin-180-binding proteins, we screened a yeast two-hybrid library using the carboxyl-terminal fragment of Densin-180 containing PDZ domain as bait, and we isolated delta-catenin/neural plakophilin-related armadillo repeat protein (NPRAP) as a Densin-180-interacting protein. delta-catenin/NPRAP, a member of the armadillo repeat family, is a nervous system-specific adherens junction protein originally discovered as an interactor with presenilin-1, a protein involved in Alzheimer's disease. Densin-180 PDZ domain binds the COOH terminus of delta-catenin/NPRAP containing the PDZ domain-binding sequence. Endogenous Densin-180 was co-immunoprecipitated with delta-catenin/NPRAP and N-cadherin. Although Densin-180 was reported to be a transmembrane protein, Densin-180 was not accessible to surface biotinylation in dissociated hippocampal neurons; hence Densin-180 may be a cytosolic protein. Densin-180 co-localized with delta-catenin/NPRAP at synapses in delta-catenin/NPRAP and may be involved in organization of the synaptic cell-cell junction through interaction with the delta-catenin/NPRAP-N-cadherin complex.