EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Inhibition and regulation of rat liver L-threonine dehydrogenase by different fatty acids and their derivatives.
PMID 11731084
Abstract
Rat liver L-threonine dehydrogenase is a mitochondrial enzyme which transforms L-threonine either into aminoacetone or into acetyl-CoA. We show that it is inhibited by several fatty acids and their derivatives: short chain fatty acids, L-2-hydroxybutyrate and D-3-hydroxybutyrate, long chain fatty acids, such as lauric acid, myristic acid, palmitic and stearic acids, bicarboxylic acids such as malonic acid and its derivatives methyl- and hydroxymalonic acids. The inhibition occurs at low and physiological concentrations of such compounds, which are normally present and metabolized in mitochondria. It presumably plays a role in the physiology of acetyl-CoA-dependent formation of fatty acids and ketobodies, in L-threonine-dependent gluconeogenesis, and in the regulation of L-threonine metabolism by L-threonine dehydrogenase and L-threonine deaminase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54917
(R)-2-Hydroxybutyric acid, ≥97.0% (T)
C4H8O3